Order Entry
Hungary
Orders LinkContactUsLinkComponent
Scratching tools, Machu
Scratching tools, Machu
Scratching tool, Basic
Catalog # SIMEVF8600
Supplier:   Industrial Physics
Scratching tools, Machu
Catalog # SIMEVF8600
Supplier:  Industrial Physics
Scratching tools, Machu
Scratching tools, Machu
Scratching tool, Basic
Price
159 000 Ft
Log in for availability
1 * 1 DB
Product Details & Documents
These scratching tools can perform a Machu corrosion test. Each tool is provided with a 1 mm width cutter to cut the coating down to the metal.
  • Ergonomic shaped grip
  • Easy to replace the cutter
  • Mandatory test in Qualicoat and QIB accredited laboratories
CC2000 has a self positioning knife bracket to expose the substrate with a perpendicular cut through the coating. CC3000 has an adjustable cutting depth and two ball bearings to guide the cut and this guarantees reproducible results.
Certifications: ISO 17872. DIN 53167, 50021, ISO 4628-8, 9227.
Documents
Specifications
  • Description:
    Scratching tool, Basic