Order Entry
Hungary
ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, CD274 (HUMAN) RECOMBINANAT PROTEIN, HUMAN CD274 RECOMBINANAT PROTEIN FUSED TO MURINE IGG2A FC AND HINGE REGION PURIFIRD FROM CHO CELLS. 1 * 25 µG
  ABNOP4938
 :  Abnova
RECOMBINANT/NATIVE PROTEIN, CD274 (HUMAN) RECOMBINANAT PROTEIN, HUMAN CD274 RECOMBINANAT PROTEIN FUSED TO MURINE IGG2A FC AND HINGE REGION PURIFIRD FROM CHO CELLS. 1 * 25 µG
  ABNOP4938
 :  Abnova
Frequently Bought Together