Order Entry
Hungary
ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, REDTAQ DNA POLYMERASE, REDTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BE EN PROVED NOT HAVING THE 3'-5' EXONUCLEASE ACTIVITY. THE ENZYME HAS PROVEN TO HAVE HIGH AMPLIFICATION YIELD, BE STABL 1 * 500 EU
Catalog # ABNOD0012
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, REDTAQ DNA POLYMERASE, REDTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BE EN PROVED NOT HAVING THE 3'-5' EXONUCLEASE ACTIVITY. THE ENZYME HAS PROVEN TO HAVE HIGH AMPLIFICATION YIELD, BE STABL 1 * 500 EU
Catalog # ABNOD0012
Supplier:  Abnova
Frequently Bought Together