Order Entry
Hungary
ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, HOTTAQ DNA POLYMERASE, HOTTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BE EN PROVED NOT HAVING THE 3' - 5'EXONUCLEASE ACTIVITY. PRIOR THE FIRST PCR STEP THE HOTTAQ DNA POLYMERASE SHOULD BE AC 1 * 500 EU
Catalog # ABNOD0011
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, HOTTAQ DNA POLYMERASE, HOTTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BE EN PROVED NOT HAVING THE 3' - 5'EXONUCLEASE ACTIVITY. PRIOR THE FIRST PCR STEP THE HOTTAQ DNA POLYMERASE SHOULD BE AC 1 * 500 EU
Catalog # ABNOD0011
Supplier:  Abnova
Frequently Bought Together