Order Entry
Hungary
Orders LinkContactUsLinkComponent
Thioisonicotinamide
Glass bottle for solids, 10 g
  CAYM13056-10
Thioisonicotinamide
  CAYM13056-10
 :  2196-13-6
Thioisonicotinamide
Glass bottle for solids, 10 g
 :  2196-13-6
45 400 Ft
1 * 10 g
 :  Pyridine-4-carbothioamide
  •  :2196-13-6
  •  :C₆H₆N₂S
  •  :MFCD00006437
  •  :138.19 g/mol
  •  :218-592-6
  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g