Accessories for empty columns HiScale™
Accessory kit for HiScale™ 16, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
Catalog # 28-9663-67
Supplier: Cytiva
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberAffinity Purification using AKTA Start.pdf Anion Exchange using AKTA Start Desalting using AKTA Start
Manual
AKTA Start Brochure
Product Literature
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Leírás:Accessory kit for HiScale™ 16, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
- Seq:37