Powder funnels, PP
150 mm, 30 mm
Catalog # 221-0317
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Reusable powder funnel, made of PP, crevice free, with wide diameter discharge spout to enable powder to be discharged without catching.
- Good chemical resistance
- Good temperature resistance
- Ideal for loading powder into drums, containers or kegs
Specifications
- Funnel O-Ø:150 mm
- Seq:02
- Stem O-Ø:30 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



