Order Entry
Hungary
Orders LinkContactUsLinkComponent
 

Caps for storage tubes, 2D barcoded, Matrix®

Caps for 0,5 ml and 1,0 ml screw top tubes, Blue
  479-2046
   Low Manufacturing Impact
Caps for storage tubes, 2D barcoded, Matrix®
  479-2046
   Low Manufacturing Impact
Caps for storage tubes, 2D barcoded, Matrix®
Caps for storage tubes, 2D barcoded, Matrix®

Caps for 0,5 ml and 1,0 ml screw top tubes, Blue

 
75 400 Ft
   
1 * 500 DB
 

Simplify sample storage with these color-coded caps. Made from Virgin class VI medical grade PP with virgin class VI medical grade silicone O-ring.

  • Free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • ScrewTop caps are compatible with storage down to −176 °C
 
  • Colour:
    Blue
  • Description:
    Caps for 0,5 ml and 1,0 ml screw top tubes
  • Seq:
    64
  • Cat. No.:
    479-2046
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact