Order Entry
Hungary
ContactUsLinkComponent
Magnetic stirrer, warming- and maintenance-free with 4 stirring points, large surface, for careful and protective mixing of cell sus pensions and culture broths, also suitable for viscous culture media. 1 * 1 items
Catalog # TMAG80004_CH
Supplier:  2MAG
Magnetic stirrer, warming- and maintenance-free with 4 stirring points, large surface, for careful and protective mixing of cell sus pensions and culture broths, also suitable for viscous culture media. 1 * 1 items
Catalog # TMAG80004_CH
Supplier:  2MAG
Frequently Bought Together