[EN]COLUMN AMIDE-80 3UM 3,2MMX1,5CM 1 * 3 DB
Catalog # TOYO0021863
Supplier: TOSOH Bioscience
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Víz, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Víz USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Etanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals