Phosphate standard solutions
Phosphate standard solution, pack of 20×2 ml ampoules (NIST), 50 mg/l as PO4, 20
Catalog # HACH17120H
Supplier: Hach
Jump to:
Product Details & Documents
Standard solution for accuracy checks in phosphate (phosphorus) determinations.
Documents
Specifications
Specifications
- Concentration:50 mg/l as PO<sub>4</sub>
- Pk:20
- Description:Phosphate standard solution, pack of 20×2 ml ampoules (NIST)