Order Entry
Hungary
Orders LinkContactUsLinkComponent
Phosphate standard solutions

Phosphate standard solutions

Phosphate standard solution, 100 ml (NIST), 15 mg/l as PO4, 100 mL
Catalog # HACH1424342
Supplier:   Hach
Phosphate standard solutions
Catalog # HACH1424342
Supplier:  Hach
CAS Number:  
Phosphate standard solutions
Phosphate standard solutions

Phosphate standard solution, 100 ml (NIST), 15 mg/l as PO4, 100 mL

CAS Number:  
Price
20 300 Ft
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solution for accuracy checks in phosphate (phosphorus) determinations.

Specifications

Specifications

  • Concentration:
    15 mg/l as PO<sub>4</sub>
  • Pk:
    100 mL
  • Description:
    Phosphate standard solution, 100 ml (NIST)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.