Order Entry
Hungary
ContactUsLinkComponent
MULTIMETER DIGITAL MULTI 3410 SET 4 : Professional digital multi meter for mobile measurement, single channel input, colour graphic display including data logger and USB interfaces. DO case set with optical IDS DO probe FDO 925, short instruction man 1 * 1 items
Catalog # 665-0218
Supplier:  WTW
MULTIMETER DIGITAL MULTI 3410 SET 4 : Professional digital multi meter for mobile measurement, single channel input, colour graphic display including data logger and USB interfaces. DO case set with optical IDS DO probe FDO 925, short instruction man 1 * 1 items
Catalog # 665-0218
Supplier:  WTW
Frequently Bought Together