Order Entry
Hungary
ContactUsLinkComponent
ChemiDoc-It 410 System. With Chemi HR camera: 12-bit monochrome, 16 File Bit Depth (A/D), digital camera and mount, 1600 x 1200 chip with binning capabilities, two stage peltier cooling to -50C from ambient, MOTORISED fixed 30mm, f/1.4 lens, UV filt 1 * 1 items
Catalog # UVPA97-0546-03
ChemiDoc-It 410 System. With Chemi HR camera: 12-bit monochrome, 16 File Bit Depth (A/D), digital camera and mount, 1600 x 1200 chip with binning capabilities, two stage peltier cooling to -50C from ambient, MOTORISED fixed 30mm, f/1.4 lens, UV filt 1 * 1 items
Catalog # UVPA97-0546-03
Frequently Bought Together