COLUMN AMIDE-80 3 UM - SILICA - 4.6MM ID X 15CM L 1 * 1 items
Catalog # 554-1603
Supplier: TOSOH Bioscience
COLUMN AMIDE-80 3 UM - SILICA - 4.6MM ID X 15CM L 1 * 1 items
Catalog # 554-1603
Supplier: TOSOH Bioscience
Frequently Bought Together
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
HPLC columns, Accucore™ aQ
Supplier: Thermo Fisher Scientific