ACID REAGENT WITH TUBING 2FT AND O-RING 1 * 1.000 mL
Catalog # ORIO182011
Supplier: Thermo Orion
Frequently Bought Together
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Maintenance solutions for electrodes
Supplier: Mettler - Toledo
REFERENCE ELECTRODE FILLING SOLUTION 1 * 5 items
Supplier: Thermo Orion
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
(±)-2-Butanol ≥99%
Supplier: Thermo Fisher Scientific
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific