Order Entry
Hungary
Orders LinkContactUsLinkComponent
Celite® 545, filter aid

Celite® 545, filter aid

1 kg
Catalog # MANA815560.1
Health
Supplier:   MACHEREY-NAGEL
Celite® 545, filter aid
Catalog # MANA815560.1
Supplier:  MACHEREY-NAGEL
CAS Number:  68855-54-9
Celite® 545, filter aid
Celite® 545, filter aid

1 kg

CAS Number:  68855-54-9
Price
16 100 Ft
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  DE, Diatomaceous earth, flux-calcined
  • CAS Number:68855-54-9
  • Formula:SiO₂
  • MDL Number:MFCD00132803
  • Molecular Weight:60.08 g/mol
  • EINECS:272-489-0
Specifications

Specifications

  • Pk:
    1 kg

Specifications