Order Entry
Hungary
ContactUsLinkComponent
to hold 96-well microtube boxes, capacity = 20 boxes in individual compartments (or (4) 96-well microtiter plates w/lids per compar tment, total of 80 w/lids) w/locking rod 18 1/2 L x 9 1/16 H x 5 1/2 W 1 * 1 items
Catalog # REVC7144
Supplier:  REVCO TECHNOLOGIES
to hold 96-well microtube boxes, capacity = 20 boxes in individual compartments (or (4) 96-well microtiter plates w/lids per compar tment, total of 80 w/lids) w/locking rod 18 1/2 L x 9 1/16 H x 5 1/2 W 1 * 1 items
Catalog # REVC7144
Supplier:  REVCO TECHNOLOGIES
Frequently Bought Together