Micro suture tying forceps, Aesculap®
Straight, 0,3 mm jaws, 150 mm
AESCFD284R
: Aesculap
These micro suture tying forceps feature a round handle.
- Type:Straight, 0,3 mm jaws
- Length:150 mm
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



