Order Entry
Hungary
Orders LinkContactUsLinkComponent
Micro suture tying forceps, Aesculap®

Micro suture tying forceps, Aesculap®

Straight, 0,3 mm jaws, 120 mm
Catalog # AESCFD280R
Supplier:   Aesculap
Micro suture tying forceps, Aesculap®
Catalog # AESCFD280R
Supplier:  Aesculap
Micro suture tying forceps, Aesculap®

Straight, 0,3 mm jaws, 120 mm

Price
107 000 Ft
Log in for availability
1 * 1 DB
Product Details & Documents

These micro suture tying forceps feature a round handle.

Specifications
  • Type:
    Straight, 0,3 mm jaws
  • Length:
    120 mm
  • Seq:
    01