Order Entry
Hungary
ContactUsLinkComponent
Dish, evaporating, material quartzglass, with spout, capacity 600ml, U-diameter 140mm, height 80mm, as per DIN 12336. 1 * 1 items
  216-1287
Dish, evaporating, material quartzglass, with spout, capacity 600ml, U-diameter 140mm, height 80mm, as per DIN 12336. 1 * 1 items
  216-1287
Frequently Bought Together