Order Entry
Hungary
Orders LinkContactUsLinkComponent
Inserts for vials, WHEATON®

Inserts for vials, WHEATON®

Limited volume insert for 1,8 ml vial, with bottom spring, 0,10 ml, Conical bottom, Glass, No
Catalog # 548-2122
Supplier:   DWK Life Sciences
Inserts for vials, WHEATON®
Catalog # 548-2122
Supplier:  DWK Life Sciences
Inserts for vials, WHEATON®
Inserts for vials, WHEATON®

Limited volume insert for 1,8 ml vial, with bottom spring, 0,10 ml, Conical bottom, Glass, No

Price
40 000 Ft
Log in for availability
1 * 100 DB
Product Details & Documents
Specifications
  • Cap included:
    No
  • Capacity:
    0,10 ml
  • Description:
    Limited volume insert for 1,8 ml vial, with bottom spring
  • Material:
    Glass
  • Base:
    Conical bottom
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.