Double bossheads
With nickel plated steel screws, Zinc die-cast, powder coated, 16,0 mm
Catalog # 241-7223
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
For stand assembly.
- Quick, easy assembly and disassembly
- Robust, durable
- Wide range of different materials available for many applications
Specifications
- Description:With nickel plated steel screws
- Material:Zinc die-cast, powder coated
- Seq:2
- For rod Ø:16,0 mm