Order Entry
Hungary
Orders LinkContactUsLinkComponent
 

Sample vials, WHEATON®

25,00 ml, 27,75×70 mm, Glass, Clear, No, 24-400
  218-2202
Sample vials, WHEATON®
  218-2202
Sample vials, WHEATON®
Sample vials, WHEATON®

25,00 ml, 27,75×70 mm, Glass, Clear, No, 24-400

 
144 000 Ft
   
1 * 200 DB
 

Borosilicate glass sample vials. Threaded without screw cap (see 548-2030 or 548-2040).

 : Packed in layers, wrapped.

 
  • Cap included:
    No
  • Cap Size:
    24-400
  • Ø×H:
    27,75×70 mm
  • Capacity:
    25,00 ml
  • Colour:
    Clear
  • Material:
    Glass
  • Seq:
    01
Ajánlott termékek

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.