Order Entry
Hungary
ContactUsLinkComponent
SONOREX DIGITAL DK 255 P, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, DIGITAL DISPLAY OF TIME, TEMPERATURE AND POWER, DEGAS-KEY, DIGI TAL TIMER 1-99 MIN AND CONTINUOUS, POWER CONTROL 10-100%, HEATING 20-80C (68-176F), INCORPORATED THERMOMETER TO SHO 1 * 1 items
Catalog # 142-1330
SONOREX DIGITAL DK 255 P, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, DIGITAL DISPLAY OF TIME, TEMPERATURE AND POWER, DEGAS-KEY, DIGI TAL TIMER 1-99 MIN AND CONTINUOUS, POWER CONTROL 10-100%, HEATING 20-80C (68-176F), INCORPORATED THERMOMETER TO SHO 1 * 1 items
Catalog # 142-1330
Frequently Bought Together