Order Entry
Hungary
ContactUsLinkComponent
Cell standard rectangular with lid 1mm pathlength, 10mm internal width, 200µl working volume, 12,5x45mm external dimensions 1 * 1 items
Catalog # 80-2003-83
Supplier:  Cytiva
Cell standard rectangular with lid 1mm pathlength, 10mm internal width, 200µl working volume, 12,5x45mm external dimensions 1 * 1 items
Catalog # 80-2003-83
Supplier:  Cytiva
Frequently Bought Together