Order Entry
Hungary
ContactUsLinkComponent
MMP-2, Human, Biotrak Assay, ELISA - 96 wells (Specific for proMMP-2: Recognizes free pro-MMP-2 and MMP-2 complexed to TIMP-2, but n ot the active form of MMP-2. No cross-reactivity with MMP-1, -3, -7, -8, -9 and MT1-MMP. Range: 1.5-24ng/ml. Sensitiv 1 * 96 Assays
Catalog # RPN2617
Supplier:  Cytiva
MMP-2, Human, Biotrak Assay, ELISA - 96 wells (Specific for proMMP-2: Recognizes free pro-MMP-2 and MMP-2 complexed to TIMP-2, but n ot the active form of MMP-2. No cross-reactivity with MMP-1, -3, -7, -8, -9 and MT1-MMP. Range: 1.5-24ng/ml. Sensitiv 1 * 96 Assays
Catalog # RPN2617
Supplier:  Cytiva
Frequently Bought Together