Phosphate standard solutions
Phosphate standard solution, 500 ml (NIST), 50 mg/l as PO4, 500 mL
Catalog # HACH17149
Supplier: Hach
Jump to:
Product Details & Documents
Standard solution for accuracy checks in phosphate (phosphorus) determinations.
Documents
Specifications
Specifications
- Concentration:50 mg/l as PO<sub>4</sub>
- Pk:500 mL
- Description:Phosphate standard solution, 500 ml (NIST)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



