Order Entry
Hungary
ContactUsLinkComponent
Filtering Bottle, GL 45
Filtering Bottle, GL 45
  519-4141
undefined
Filtering Bottle, GL 45
  519-4141
  • Capacity:
    2000 ml
  • Description:
    Filtering bottle, GL 45
  • Pk:
    1
ROBU® Filtering bottle with GL 45 thread, low pressure resistant with screw-on plastic hose, protective coating, sterilisable
  • High-quality GL 45 screw cap
  • Includes aperture for easy access to contents
  • Complete set with durable silicone washer for a secure seal
  • Made from chemical-resistant materials
  • Provides a tight and leak-proof fit
  • Ensures optimal protection against contamination
  • Reusable and easy to clean
The ROBU filtering bottle, GL 45, is designed for efficient filtration and collection of liquids in laboratory applications. Made from high-quality borosilicate glass, this bottle features a GL 45 threaded neck for secure sealing with compatible caps. The integrated design allows for easy filtration, ideal for applications such as sample preparation, chemical analysis, and microbiological studies. The durable construction ensures resistance to chemicals, high temperatures, and mechanical stress. The ROBU filtering bottle provides a reliable, leak-proof solution for liquid filtration, offering consistent performance in both routine and specialised laboratory tasks.
Caps