Isofield Blazer LT Sterile Latex Cleanroom Gloves
Supplier: ISOFIELD IRELAND
New Product
Certificates
About this item
Isofield Blazer LT sterile latex cleanroom gloves are suitable for use in GMP Grade A and B, ISO Class 4 cleanrooms and above.
Hand-specific
- Natural colour
- Textured palm and fingers
- Beaded cuff
- Low endotoxin levels
- Protein level less than 50 μg/g
- Gamma irradiated, minimum 25 kGy
- Sterility Assurance Level 10⁻⁶
- IPA resistant ink pouches
This ultra-clean latex glove is triple washed in deionized water and its surface is smooth enough for double-donning whilst also offering a textured palm and fingers for added grip. Every Blazer LT lot is tested for endotoxins and has very low levels of detectable protein (less than 50 µg/g).
Certifications:
EN ISO 21420, EN ISO 374-1, EN ISO 374-2, EN ISO 374-4, EN ISO 374-5, EN ISO 16523, EN ISO 16604.
Packaging:
Gloves are packaged 1 pair per inner PE wallet, 1 PE wallet per sealed PE pouch, 10 pouches per PE bag, 20 sealed PE bags per lined carton (200 pairs).
Specifications
- Catalog No:
- ISOF1063070LT
- ISOF1063075LT
- ISOF1063080LT
- ISOF1063010LT
- ISOF1063060LT
- ISOF1063085LT
- ISOF1063065LT
- ISOF1063090LT
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded