Order Entry
Hungary
ContactUsLinkComponent
 
NANOCOLOR® Photometric Standard Test Kit for Silica
  634-8130
 :  
undefined
NANOCOLOR® Photometric Standard Test Kit for Silica
  634-8130
 :  

 

  • Chemical method:
    Silicomolybdenum blue
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Silica, test no. 1-48
  • Dimensions:
    160×110×130 mm (including packaging)
  • Model:
    MANA91848
  • Platform:
    NANOCOLOR® standard tests
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    36
  • Weight:
    436 g (including packaging)
  • For unit:
    UV/VIS II; VIS II; Advance; 500 D
  • Tests per kit:
    250
  • Range:
    0,01 - 5,00 mg/L (Si); 0,02 - 10,0 mg/L (SiO₂)

 

 

Photometric test kit for the determination of silica in water samples.

  • Ready-to-use reagents for up to 500 determinations
  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II

 : Rugged box with reagents for 250 tests.