Order Entry
Hungary
ContactUsLinkComponent
 
NANOCOLOR® Photometric Standard Test Kit for Chlorine Dioxide
  MANA918163
 :  
undefined
NANOCOLOR® Photometric Standard Test Kit for Chlorine Dioxide
  MANA918163
 :  

 

  • Chemical method:
    DPD
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Chlorine dioxide, test no. 1-163
  • Dimensions:
    160×110×130 mm (Including packaging)
  • Model:
    MANA918163
  • Platform:
    NANOCOLOR® standard tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    18
  • Weight:
    471 g (Including packaging)
  • For unit:
    UV/VIS II;VIS II;Advance;500 D
  • Tests per kit:
    50
  • Range:
    0,04 - 4,00 mg/L

 

 

Photometric test kit for the determination of chlorine dioxide in water samples.

  • Ready-to-use reagents for up to 500 determinations

  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sea water analysis: Yes

Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II

Sludge reagent set: No

Digestion set: No

NanOx Metal: No

NanOx N: No

 : Rugged box with reagents for 50 tests.