b.safe Screw Caps
Catalog # BOHLM575-01
Supplier: Bohlender
New Product
Specifications
- Thread:GL 45
- Material:PP
Specifications
About this item
Screw cap made of PP with handy knurl and internal PTFE/silicone gasket. After assembly, the product is only exposed to PTFE.
Made of PP
- Very good chemical resistance
- Equipped with PTFE/silicone seal
Screw caps are designed for closing unused connections on a b.safe Distributor for flushing bottles, or also to close eluent bottles during the flushing process.