Order Entry
Hungary
ContactUsLinkComponent
Mouse Alpha-1-Microglobulin ELISA Kit
Mouse Alpha-1-Microglobulin ELISA Kit
Catalog # ABCAAB316256-96TST
Supplier:  Abcam
New Product
CAS Number:  
undefined
Mouse Alpha-1-Microglobulin ELISA Kit
Catalog # ABCAAB316256-96TST
Supplier:  Abcam
New Product
CAS Number:  

Specifications

  • Description:
    Mouse Alpha-1-Microglobulin ELISA kit
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse Alpha-1-Microglobulin ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Alpha-1-Microglobulin with a sensitivity of 1,932 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.