Mouse/Rat Cytochrome C ELISA Kit
Katalógus# ABCAAB210575-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse/Rat Cytochrome C ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse/Rat Cytochrome C ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse/Rat Cytochrome C with a sensitivity of 44 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader. - Cited in over 5 citations.