Human Superoxide Dismutase 1 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB253866-10X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Superoxide Dismutase 1
- Description:Human Superoxide Dismutase 1 antibody pair - BSA and azide free
- Detection range:0,31 - 20 ng/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Human Superoxide Dismutase 1 antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Superoxide Dismutase 1.