Monkey MIP-1 alpha/CCL3 ELISA Kit
ABCAAB289698-1X96
: Abcam
:
- Description:Monkey MIP-1 alpha/CCL3 ELISA kit
- Tests per kit:96 tests
Monkey MIP-1 alpha/CCL3 ELISA Kit is a single-wash 90-min Simplestep used to quantify Monkey MIP-1 alpha/CCL3 with a sensitivity of 8,7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.