Mouse XCL1 ELISA Kit (Lymphotactin)
Catalog # ABCAAB269540-1X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:XCL1
- Description:Mouse XCL1 ELISA kit (Lymphotactin)
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Detection range:18,75 - 1200 pg/ml
- Storage temperature:4 °C
- Sensitivity:6,26 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse XCL1 ELISA kit (Lymphotactin) is a single-wash 90-min Simplestep used to quantify Mouse XCL1 (Lymphotactin) with a sensitivity of 6.26 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.