Monkey MMP3 ELISA Kit
ABCAAB315314-96TST
: Abcam
New Product
:
- Description:Monkey MMP3 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Monkey MMP3 ELISA kit is a single-wash 90-min Simplestep used to quantify Monkey MMP3 with a sensitivity of 10,45 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Validated on a number of sample types including cerebrospinal fluid (CSF).