Mouse Met ELISA Kit (c-Met)
ABCAAB275107-1X96
: Abcam
:
- Description:Mouse Met ELISA Kit (c-Met)
- Tests per kit:96 tests
Mouse Met ELISA Kit (c-Met) is a single-wash 90-min Simplestep used to quantify Mouse Met (c-Met) with a sensitivity of 10 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.