Cleaved Tau ELISA Kit (Human Asp738/Mouse Asp713)
Catalog # ABCAAB269557-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Cleaved Tau ELISA kit (Human Asp738/Mouse Asp713)
Specifications
About this item
Cleaved Tau ELISA kit (Human Asp738/Mouse Asp713) is a single-wash 90-min Simplestep used to quantify Cleaved Tau (Human Asp738/Mouse Asp713) with a sensitivity of 0,075 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.