Order Entry
Hungary
ContactUsLinkComponent
Mouse MAG/GMA ELISA Kit
Mouse MAG/GMA ELISA Kit
Catalog # ABCAAB289652-1X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse MAG/GMA ELISA Kit
Catalog # ABCAAB289652-1X96
Supplier:  Abcam
CAS Number:  
About this item
Mouse MAG/GMA ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse MAG/GMA with a sensitivity of 9.285 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.