Order Entry
Hungary
ContactUsLinkComponent
Mouse PRAS40 ELISA Kit
Mouse PRAS40 ELISA Kit
Catalog # ABCAAB210580-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse PRAS40 ELISA Kit
Catalog # ABCAAB210580-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse PRAS40 ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse PRAS40 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse PRAS40 with a sensitivity of 23,5 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.
Certifications: RUO