Human Interferon gamma Antibody Pair - BSA and Azide Free
Catalog # ABCAAB290008-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Interferon gamma antibody pair - BSA and Azide Free
- Tests per kit:10×96 tests
Specifications
About this item
Human Interferon gamma Antibody pair - BSA and Azide Free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Interferon gamma.