Monkey Eotaxin ELISA Kit
About this item
Monkey Eotaxin ELISA kit is a single-wash 90-min Simplestep used to quantify Monkey Eotaxin with a sensitivity of 12.913 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.
2 Options Available Below
Product Details & Documents
Monkey Eotaxin ELISA kit is a single-wash 90-min Simplestep used to quantify Monkey Eotaxin with a sensitivity of 12.913 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ABCAAB317549-96TST
- ABCAAB317549-1X384
- Target protein:
- Eotaxin
- Eotaxin
- Primary antibody reactivity:
- Monkey
- Monkey
- Assay Type:
- Sandwich
- Sandwich
- Format:
- Pre-coated
- Pre-coated
- Assay duration:
- One step
- One step
- Sensitivity:
- 12,913 pg/ml
- 12,913 pg/ml
- Detection range:
- 39,063 - 2500 pg/ml
- 39,063 - 2500 pg/ml
- Detection method:
- Colorimetric
- Colorimetric
- Sample type:
- Serum, EDTA Plasma, Citrate plasma
- Serum, EDTA Plasma, Citrate plasma
- Time to Results:
- 1 hr 30 min
- 1 hr 30 min
- Regulatory status:
- RUO
- RUO
- Storage temperature:
- 4 °C
- 4 °C
Specifications
Product Family Options
Product Information
- Tests per kitAvailabilityPrice
- ABCAAB317549-96TSTMonkey Eotaxin ELISA KitSpecifications:New Product
More
Specifications
Cat. No.ABCAAB317549-96TSTTarget proteinEotaxinPrimary antibody reactivityMonkeyAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity12,913 pg/mlDetection range39,063 - 2500 pg/mlDetection methodColorimetricSample typeSerum, EDTA Plasma, Citrate plasmaTime to Results1 hr 30 minRegulatory statusRUOStorage temperature4 °CNew Product - ABCAAB317549-1X384Monkey Eotaxin ELISA KitSpecifications:New Product
More
Specifications
Cat. No.ABCAAB317549-1X384Target proteinEotaxinPrimary antibody reactivityMonkeyAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity12,913 pg/mlDetection range39,063 - 2500 pg/mlDetection methodColorimetricSample typeSerum, EDTA Plasma, Citrate plasmaTime to Results1 hr 30 minRegulatory statusRUOStorage temperature4 °CNew Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



