Mouse ST2 (IL1 RL1) ELISA Kit
Catalog # ABCAAB309171-96TST
Gyártó: Abcam
CAS szám:
Specifikációk
- Description:Mouse ST2 (IL1 RL1) ELISA Kit
- Tests per kit:96 tests
Specifikációk
Termék információk
Mouse ST2 (IL1 RL1) ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse ST2 (IL1 RL1) with a sensitivity of 11,54 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.