Human TFF3 Antibody Pair - BSA and Azide free
Catalog # ABCAAB244090-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human TFF3 antibody pair - BSA and Azide free
- Tests per kit:10×96 tests
Specifications
About this item
Human TFF3 Antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human TFF3.