Mouse IFN beta ELISA Kit
Catalog # ABCAAB252363-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse IFN beta ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse IFN beta ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse IFN beta with a sensitivity of 28 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput. - Cited in over 15 citations.