Mouse Flt3 ligand ELISA Kit (Flt3L)
ABCAAB275551-1X96
: Abcam
:
- Description:Mouse Flt3 ligand ELISA kit (Flt3L)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Mouse Flt3 ligand ELISA kit (Flt3L) is a single-wash 90-min Simplestep used to quantify Mouse Flt3 ligand (Flt3L) with a sensitivity of 3,9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.