Human DAP12 (pY91) + DAP12 Total ELISA Kit
Catalog # ABCAAB316136-96TST
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Human DAP12 (pY91) + DAP12 Total ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human DAP12 (pY91) + DAP12 Total ELISA Kit is a single-wash 90-min Simplestep used to detect Human DAP12 (pY91) + DAP12 Total. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Semi-quantitative Sandwich ELISA - 450 nm readout : works on any standard plate reader.